Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Jars with overlapping lid

Jars with overlapping lid

25 ml, 40 mm, 32 mm
Catalog # SEMA3010
Supplier:   SEMADENI
Jars with overlapping lid
Catalog # SEMA3010
Supplier:  SEMADENI
Jars with overlapping lid
Jars with overlapping lid

25 ml, 40 mm, 32 mm

Price
SFr. 24.90
Log in for availability
1 * 100 ST
Product Details & Documents

Jars, made of PP, with overlapping lid.

  • Lids can be labelled
  • Suitable for ointments, powders, and granules
  • Good chemical and temperature resistance
Specifications
  • Ø est:
    40 mm
  • Capacità:
    25 ml
  • Altezza:
    32 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.