Containers, rectangular
8,5 l, 445×345×90 mm, bianco, HDPE
SEMA1295
: SEMADENI
HDPE or PP, white, stackable.
These are ideal containers with side handles and edge for gripping to handle solid samples. Containers over 225 mm height are not suitable for liquids.
- Capacità:8,5 l
- Colore:bianco
- Materiale:HDPE
- Seq:01
- L×P×A:445×345×90 mm