Jars with screw cap
2000 ml, 141 mm, 164 mm
Codice catalogo SEMA5750
Nome del fornitore: SEMADENI
Vai a
Product Details & Documents
Jars, made of PP, with tight screw cap.
Suitable for powders, granules, pastes, ointments, waxes, cosmetics and dermatology products.
Specifiche
- Ø est:141 mm
- Capacità:2000 ml
- Altezza:164 mm



