Jars with overlapping lid
68 mm, 16 mm
Codice catalogo SEMA1712
Nome del fornitore: SEMADENI
Vai a
Product Details & Documents
Jars, made of PS, clear, with closing lid.
- Good chemical resistance
- Easy to clean
Suitable for ointments, powders, and granules.
Specifiche
- Ø est:68 mm
- Altezza:16 mm
- Seq:11
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



