Tubing ptfe 0.8x1.6mm for the highest standards, good chemical and temperature resistant from -220 c to +260 c. the ptfe tube ha s a low coefficient of friction, is non-stick (easy to clean) and uv-resistant. 1 * 1 m
Codice catalogo SEMA5699
Nome del fornitore: SEMADENI
Frequently Bought Together