Jars with overlapping lid
25 mm, 25 mm
Codice catalogo SEMA3390
Nome del fornitore: SEMADENI
Vai a
Product Details & Documents
Jars, made of PS, clear, with closing lid.
- Good chemical resistance
- Easy to clean
Suitable for ointments, powders, and granules.
Specifiche
- Ø est:25 mm
- Altezza:25 mm
- Seq:10