Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Pallets, gas cylinders

Transport pallet for max. 8× Ø 250 mm gas cylinders, load capacity 700 kg, 1200×860×1000 mm
  DUEH7.00.810
 :   DUPERTHAL
Pallets, gas cylinders
  DUEH7.00.810
 :  DUPERTHAL
Pallets, gas cylinders
Pallets, gas cylinders

Transport pallet for max. 8× Ø 250 mm gas cylinders, load capacity 700 kg, 1200×860×1000 mm

 
SFr. 579.00
   
1 * 1 ST
 

These pallets are used for the safe, economic and effortless transport of 1 to 8 gas cylinders with hot-dip galvanised surface.

  • Sturdy base frame with fortlift pockets
  • Gas cylinders can be secured individually with chains
  • Lateral tilt limiter
  • Hooks for hoses and screw caps

Inner dimensions of fortlift pockets (W×H): 170×70 mm.

Inner distance between pockets: 510 mm.

 : According to EN ISO 1461

 
  • Descrizione:
    Transport pallet for max. 8× Ø 250 mm gas cylinders, load capacity 700 kg
  • L×P×A:
    1200×860×1000 mm