DirectPCR® DNA extraction system
DirectPCR®-Cell, optimised for lysis of mammalian cells, 100 ml, 100 mL
Codice catalogo 732-3260
Nome del fornitore: VIAGEN BIOTECH
DirectPCR® DNA extraction system
Codice catalogo 732-3260
Nome del fornitore: VIAGEN BIOTECH
Numero CAS:
Vai a
Product Details & Documents
DirectPCR® DNA Lysis Reagents offer a quick and cost-effective 'single-tube' system for the rapid preparation of PCR template DNA from mouse tails, ears, embryonic material or mammalian cells.
- 'Single-tube' system for reduced number of pipetting steps
- Suitable for the simultaneous processing of multiple samples
- No need for organic extractions or time-consuming alcohol precipitations
- Reliable and absolutely reproducible procedure
- High yields
- Lysate can directly be used for genotyping PCRs
NOTE: 1 µl lysate is sufficient for subsequent genotyping PCR reactions.
Specifiche
Specifiche
- Conf:100 mL
- Descrizione:DirectPCR®-Cell, optimised for lysis of mammalian cells, 100 ml