Orders LinkContactUsLinkComponent
HPLC columns, C18
HPLC columns, C18
Fortis C18 HPLC columns, 1,7 µm, 3,00 mm, 50 mm
Codice catalogo FORTF18-030301
Nome del fornitore:   FORTIS TECHNOLOGIES
HPLC columns, C18
Codice catalogo FORTF18-030301
Nome del fornitore:  FORTIS TECHNOLOGIES
HPLC columns, C18
Fortis C18 HPLC columns, 1,7 µm, 3,00 mm, 50 mm
Price
Richiedi prezzo
Log in per disponibilità
1 * 1 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Fortis C18 is a pure silica based stationalry phase with unique high and low pH performance.
  • pH range 1 to 12
  • Provide superior peak shapes
  • Fully scalable - UHPLC to prep
Specifiche
  • Ø int.:
    3,00 mm
  • Dimensioni particelle:
    1,7 µm
  • Descrizione:
    Fortis C18 HPLC columns
  • Lunghezza:
    50 mm
  • Seq:
    07
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.