Orders LinkContactUsLinkComponent
HPLC columns, C18
Fortis C18 HPLC columns, 2,5 µm, 2,10 mm, 50 mm
Catalog # FORTF18-020302
Supplier:   FORTIS TECHNOLOGIES
HPLC columns, C18
Catalog # FORTF18-020302
HPLC columns, C18
Fortis C18 HPLC columns, 2,5 µm, 2,10 mm, 50 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Fortis C18 is a pure silica based stationalry phase with unique high and low pH performance.
  • pH range 1 to 12
  • Provide superior peak shapes
  • Fully scalable - UHPLC to prep
Specifications
  • Ø int.:
    2,10 mm
  • Dimensioni particelle:
    2,5 µm
  • Descrizione:
    Fortis C18 HPLC columns
  • Lunghezza:
    50 mm
  • Seq:
    13
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.