Hydrophobic interaction chromatography columns, TSK-GEL®
Prep guardgel kit, for P/N 07656, Precolonna, Phenyl-5PW, 20 µm
Codice catalogo TOYO0016095
Nome del fornitore: TOSOH Bioscience
Hydrophobic interaction chromatography columns, TSK-GEL®
Codice catalogo TOYO0016095
Nome del fornitore: TOSOH Bioscience
Vai a
Product Details & Documents
These are polymethacrylate-based columns with a choice of three ligands (ether, phenyl or butyl) with varied hydrophobicities from low to high. Suitable for LC-MS applications.
- Rigid polymeric base resin
- Eliminates undesirable interactions with column hardware
- Wide pH range (2 to 12) enables robust cleaning options
Certificates
Cerca per numero di lottoSpecifiche
- Formato della colonna:Precolonna
- Dimensioni particelle:20 µm
- Descrizione:Prep guardgel kit, for P/N 07656
- Fase:Phenyl-5PW