Precision tweezers, resistant, DURAX™
Straight, extra long, very fine, SS Durax™, 135 mm
Codice catalogo OUTISS-DURAX
Nome del fornitore: OUTILS RUBIS
Precision tweezers, resistant, DURAX™
Codice catalogo OUTISS-DURAX
Nome del fornitore: OUTILS RUBIS
Vai a
Product Details & Documents
DURAX™ (Martensitic steel).
- Hardened up to 616 Vickers: becomes six times harder than standard steel
- High resistance against wear and tear: Longer service life than ordinary tweezers
Specifiche
- Tipo:SS Durax™
- Descrizione:Straight, extra long, very fine
- Lunghezza:135 mm
- Seq:10