Orders LinkContactUsLinkComponent
Precision tweezers, resistant, DURAX™
Precision tweezers, resistant, DURAX™
Straight, very fine, rounded edges, 3 Durax™, 120 mm
Catalog # OUTI3-DURAX
Supplier:   OUTILS RUBIS
Precision tweezers, resistant, DURAX™
Catalog # OUTI3-DURAX
Supplier:  OUTILS RUBIS
Precision tweezers, resistant, DURAX™
Precision tweezers, resistant, DURAX™
Straight, very fine, rounded edges, 3 Durax™, 120 mm
Price
SFr. 48.70
Log in for availability
1 * 1 ST
Product Details & Documents
DURAX™ (Martensitic steel).
  • Hardened up to 616 Vickers: becomes six times harder than standard steel
  • High resistance against wear and tear: Longer service life than ordinary tweezers
Specifications
  • Tipo:
    3 Durax™
  • Descrizione:
    Straight, very fine, rounded edges
  • Lunghezza:
    120 mm
  • Seq:
    05