Precision tweezers, resistant, DURAX™
Straight, very fine, edges on the outside rounded off, 1 Durax™, 120 mm
Codice catalogo OUTI1-DURAX
Nome del fornitore: OUTILS RUBIS
Precision tweezers, resistant, DURAX™
Codice catalogo OUTI1-DURAX
Nome del fornitore: OUTILS RUBIS
Vai a
Product Details & Documents
DURAX™ (Martensitic steel).
- Hardened up to 616 Vickers: becomes six times harder than standard steel
- High resistance against wear and tear: Longer service life than ordinary tweezers
Specifiche
- Tipo:1 Durax™
- Descrizione:Straight, very fine, edges on the outside rounded off
- Lunghezza:120 mm
- Seq:03