Orders LinkContactUsLinkComponent
Precision tweezers, resistant, DURAX™
Precision tweezers, resistant, DURAX™
Straight, very fine, edges on the outside rounded off, 1 Durax™, 120 mm
Codice catalogo OUTI1-DURAX
Nome del fornitore:   OUTILS RUBIS
Precision tweezers, resistant, DURAX™
Codice catalogo OUTI1-DURAX
Nome del fornitore:  OUTILS RUBIS
Precision tweezers, resistant, DURAX™
Precision tweezers, resistant, DURAX™
Straight, very fine, edges on the outside rounded off, 1 Durax™, 120 mm
Price
SFr. 42.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
DURAX™ (Martensitic steel).
  • Hardened up to 616 Vickers: becomes six times harder than standard steel
  • High resistance against wear and tear: Longer service life than ordinary tweezers
Specifiche
  • Tipo:
    1 Durax™
  • Descrizione:
    Straight, very fine, edges on the outside rounded off
  • Lunghezza:
    120 mm
  • Seq:
    03