Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Friedman-Pearson rongeurs

Friedman-Pearson rongeurs, 1 mm cup diameter, Straight, 140 mm
Codice catalogo FSCI16020-14
Marketsource
Nome del fornitore:   FINE SCIENCE TOOLS
Friedman-Pearson rongeurs
Codice catalogo FSCI16020-14
Nome del fornitore:  FINE SCIENCE TOOLS
Marketsource
Friedman-Pearson rongeurs
Friedman-Pearson rongeurs

Friedman-Pearson rongeurs, 1 mm cup diameter, Straight, 140 mm

Price
254.22 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents

These rongeurs are made of stainless steel.

  • Single joint action
Specifiche
  • Tipo:
    Straight
  • Descrizione:
    Friedman-Pearson rongeurs, 1 mm cup diameter
  • Lunghezza:
    140 mm
  • Seq:
    04