Orders LinkContactUsLinkComponent
Spring scissors, 8 mm cutting edge
Sharp, straight tips, 100 mm, 8 mm, 0,15 mm
  FSCI15024-10
Marketsource
Spring scissors, 8 mm cutting edge
  FSCI15024-10
Marketsource
Spring scissors, 8 mm cutting edge
Spring scissors, 8 mm cutting edge
Sharp, straight tips, 100 mm, 8 mm, 0,15 mm
SFr. 178.64
1 * 1 ST
These scissors are made of stainless steel.
  • Tipo:
    Sharp, straight tips
  • Lunghezza:
    100 mm
  • Seq:
    03
  • Tip size:
    0,15 mm
  • Lunghezza lama:
    8 mm