Surgical scissors, blunt
Blunt/blunt, curved tips, 185 mm, 57 mm
FSCI14003-18
Marketsource
These scissors are made of stainless steel.
- Tipo:Blunt/blunt, curved tips
- Lunghezza:185 mm
- Seq:09
- Lunghezza lama:57 mm
These scissors are made of stainless steel.