Surgical scissors, blunt
Blunt/blunt, straight tips, 145 mm, 47 mm
Codice catalogo FSCI14000-14
Marketsource
Nome del fornitore: FINE SCIENCE TOOLS
Surgical scissors, blunt
Codice catalogo FSCI14000-14
Nome del fornitore: FINE SCIENCE TOOLS
Marketsource
Vai a
Product Details & Documents
These scissors are made of stainless steel.
Specifiche
- Tipo:Blunt/blunt, straight tips
- Lunghezza:145 mm
- Seq:06
- Lunghezza lama:47 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



