Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Fine scissors, ToughCut®

Fine scissors, ToughCut®

Sharp/sharp, straight tips, Toughcut®, 115 mm, 25 mm, +
N° de catalogue FSCI14058-11
Marketsource
Fournisseur:   FINE SCIENCE TOOLS
Fine scissors, ToughCut®
N° de catalogue FSCI14058-11
Fournisseur:  FINE SCIENCE TOOLS
Marketsource
Fine scissors, ToughCut®
Fine scissors, ToughCut®

Sharp/sharp, straight tips, Toughcut®, 115 mm, 25 mm, +

Prix
SFr. 53.59
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

These scissors are made of stainless steel.

Spécifications
  • Tipo:
    Sharp/sharp, straight tips, Toughcut®
  • Lunghezza:
    115 mm
  • Seq:
    04
  • Serrated:
    +
  • Lunghezza lama:
    25 mm