Metzenbaum scissors
Blunt/blunt, curved tips, Toughcut® , 180 mm, 29 mm, +
N° de catalogue FSCI14117-18
Marketsource
Fournisseur: FINE SCIENCE TOOLS
Aller à
Product Details & Documents
These scissors are made of stainless steel.
Spécifications
- Tipo:Blunt/blunt, curved tips, Toughcut®
- Lunghezza:180 mm
- Seq:02
- Serrated:+
- Lunghezza lama:29 mm