Sample cups for analysers, ratiolab®
For Technicon T2 analysers, 3,5 ml
Codice catalogo RATI5700120
Nome del fornitore: RATIOLAB
Vai a
Product Details & Documents
Made of clear PS.
- Good chemical resistance
- Good temperature resistance
Analysis beakers offer the highest precision for use with robotic analytical systems.
Specifiche
- Capacità:3,5 ml
- Descrizione:For Technicon T2 analysers
- Seq:04