Fmoc-Pro-OH
Glass bottle for solids, 500 g
Catalog # BACE4003150.0500
Marketsource
Supplier: BACHEM BIOCHEMICA
Fmoc-Pro-OH
Catalog # BACE4003150.0500
Supplier: BACHEM BIOCHEMICA
Marketsource
CAS Number: 71989-31-6
Jump to:
Product Details & Documents
Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-proline, N-Fmoc-L-proline
- CAS Number:71989-31-6
- Formula:C₂₀H₁₉NO₄
- MDL Number:MFCD00037122
- Molecular Weight:337.38 g/mol
- EINECS:276-259-0
Specifications
Specifications
- Tipo di conf.:Glass bottle for solids
- Conf:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



