n-Dodecil-β-D-maltoside
Glass bottle for solids, 1 g
Catalog # BACE4061027.0001
Marketsource
Supplier: BACHEM BIOCHEMICA
n-Dodecil-β-D-maltoside
Catalog # BACE4061027.0001
Supplier: BACHEM BIOCHEMICA
Marketsource
CAS Number: 69227-93-6
Jump to:
Product Details & Documents
Synonyms: DDM, Lauryl-β-D-maltoside, Dodecyl-β-D-maltopyranoside, (2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, N-Dodecyl-Beta-Maltoside
- CAS Number:69227-93-6
- Formula:C₂₄H₄₆O₁₁
- MDL Number:MFCD00043012
- Molecular Weight:510.6 g/mol
- Storage Temperature:Congelamento
Specifications
Specifications
- Tipo di conf.:Glass bottle for solids
- Conf:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



