Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Funnels with short stem

Funnels with short stem

20 mm, 260 mm
Codice catalogo HUEN954100
Nome del fornitore:   hünersdorff GmbH
Funnels with short stem
Codice catalogo HUEN954100
Nome del fornitore:  hünersdorff GmbH
Funnels with short stem
Funnels with short stem

20 mm, 260 mm

Price
SFr. 45.60
Log in per disponibilità
1 * 6 ST
Product Details & Documents

These food safe HDPE funnels are acid and chemical resistant and are ideal for use in the laboratory, workshop and household purposes.

  • Food-safe
  • Good chemical resistance
  • Good temperature resistance
Specifiche
  • Ø esterno imbuto:
    260 mm
  • Seq:
    05
  • Ø esterno stelo:
    20 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.