Orders LinkContactUsLinkComponent
Maxima™ First strand cDNA synthesis kit
Maxima™ First strand cDNA synthesis kit
Maxima® First Strand cDNA Synthesis Kit for RT-qPCR, 50 reactions, 1 KIT
Codice catalogo K1641
Nome del fornitore:   Thermo Fisher Scientific
Maxima™ First strand cDNA synthesis kit
Codice catalogo K1641
Nome del fornitore:  Thermo Fisher Scientific
Numero CAS:  
Maxima™ First strand cDNA synthesis kit
Maxima™ First strand cDNA synthesis kit
Maxima® First Strand cDNA Synthesis Kit for RT-qPCR, 50 reactions, 1 KIT
Numero CAS:  
Price
SFr. 700.00
Log in per disponibilità
1 * 1 KIT
Product Details & Documents
The Maxima™ First Strand cDNA Synthesis Kit is a convenient system optimised for use as a first step in two step RT-qPCR. The kit includes the Maxima™ Reverse Transcriptase, which is an advanced enzyme derived by in vitro evolution of M-MuLV RT for RT-qPCR application. The enzyme features high thermostability, robustness and increased synthesis rates in RT-qPCR.
  • Increased reaction temperatures – up to 60 °C
  • Premixed components for convenient use in RT-qPCR
  • Increased synthesis rate – complete cDNA synthesis in 15 minutes
  • High sensitivity and specificity
The Maxima™ First Strand cDNA Synthesis Kit is capable of cDNA synthesis at elevated temperatures (50 to 60 °C) and completion of the reverse transcription reaction in 15 minutes. In addition, the kit offers highly convenient composition, where Maxima™ Reverse Transcriptase and RiboLock™ RNase Inhibitor are premixed, while all other reaction components: reaction buffer, dNTPs and primers (random hexamers and Oligo(dT)₁₈) are premixed in another tube as a 5X reaction mix.
Informazioni per l'ordine: Kit includes Maxima™ enzyme mix, 5X reaction mix, nuclease-free water, and detailed protocol.
Specifiche
Specifiche
  • Conf:
    1 KIT
  • Descrizione:
    Maxima® First Strand cDNA Synthesis Kit for RT-qPCR, 50 reactions