Orders LinkContactUsLinkComponent
Safety spectacles, Slam
Safety spectacles, Slam
Anti-UV, anti-scratch, anti-fog, anti-static, nero, Clear PC
Codice catalogo 111-1014
Nome del fornitore:   Bollé Safety
Safety spectacles, Slam
Codice catalogo 111-1014
Nome del fornitore:  Bollé Safety
Safety spectacles, Slam
Safety spectacles, Slam
Anti-UV, anti-scratch, anti-fog, anti-static, nero, Clear PC
Price
9.50 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Modern design spectacles with extra-easy fitting and comfort.
  • Panoramic visual field
  • Anti-scratch, anti-fog, anti-static coatings
  • Ultra enveloping
  • Comfort temples
Weight: 23 g.
Certifications: EN 166, EN 170 (clear lens), EN 172 (smoke lens).
Delivery information: Supplied with adjustable cord.
Specifiche
  • Tipo:
    Anti-UV, anti-scratch, anti-fog, anti-static
  • Colore:
    nero
  • Seq:
    02
  • Materiale lente:
    Clear PC