Safety spectacles, Slam
Anti-UV, anti-scratch, anti-fog, anti-static, nero, Clear PC
Codice catalogo 111-1014
Nome del fornitore: Bollé Safety
Vai a
Product Details & Documents
Modern design spectacles with extra-easy fitting and comfort.
- Panoramic visual field
- Anti-scratch, anti-fog, anti-static coatings
- Ultra enveloping
- Comfort temples
Weight: 23 g.
Certifications: EN 166, EN 170 (clear lens), EN 172 (smoke lens).
Delivery information: Supplied with adjustable cord.
Specifiche
- Tipo:Anti-UV, anti-scratch, anti-fog, anti-static
- Colore:nero
- Seq:02
- Materiale lente:Clear PC