2-Chlorophenylboronic acid
Glass bottle for solids, 25 g
Catalog # COBBBB-2259-25G
Marketsource
Supplier: COMBI-BLOCKS
2-Chlorophenylboronic acid
Catalog # COBBBB-2259-25G
Supplier: COMBI-BLOCKS
Marketsource
CAS Number: 3900-89-8
Jump to:
Product Details & Documents
Synonyms: (2-Chlorophenyl)metaboric acid, Dihydroxy(2-chlorophenyl)borane, (2-Chlorophenyl)boronic acid, 2-Chlorobenzeneboronic acid
- CAS Number:3900-89-8
- Formula:C₆H₆BClO₂
- MDL Number:MFCD00674012
- Molecular Weight:156.38 g/mol
Specifications
Specifications
- Tipo di conf.:Glass bottle for solids
- Conf:25 g



