4-Methoxybenzonitrile
Bottiglia di vetro, 25 g
Catalog # COBBCA-4623-25G
Marketsource
Supplier: COMBI-BLOCKS
4-Methoxybenzonitrile
Catalog # COBBCA-4623-25G
Supplier: COMBI-BLOCKS
Marketsource
CAS Number: 874-90-8
Jump to:
Product Details & Documents
Synonyms: 4-Cyanoanisol, Anisonitrile, p-Cyanoanisol, p-Methoxybenzonitrile
- CAS Number:874-90-8
- Formula:C₈H₇NO
- MDL Number:MFCD00001818
- Molecular Weight:133.15 g/mol
- Boiling Pt:256 °C (1013 hPa)
- Melting Pt:57…60 °C
- Storage Temperature:Ambiente
- EINECS:212-871-6
- UN:3439
- ADR:6.1,III
Specifications
Specifications
- Tipo di conf.:Bottiglia di vetro
- Conf:25 g



