5-Fluoro-2-nitroanisole
Glass bottle for solids, 5 g
Catalog # COBBAN-1018-5G
Marketsource
Supplier: COMBI-BLOCKS
5-Fluoro-2-nitroanisole
Catalog # COBBAN-1018-5G
Supplier: COMBI-BLOCKS
Marketsource
CAS Number: 448-19-1
Jump to:
Product Details & Documents
Synonyms: 4-Fluoro-2-methoxy-1-nitrobenzene
- CAS Number:448-19-1
- Formula:C₇H₆FNO₃
- MDL Number:MFCD00077541
- Molecular Weight:171.13 g/mol
- Storage Temperature:Ambiente
Specifications
Specifications
- Tipo di conf.:Glass bottle for solids
- Conf:5 g



