4-Bromoindole
Glass bottle for solids, 1 g
Catalog # COBBIN-0043-1G
Marketsource
Supplier: COMBI-BLOCKS
Jump to:
Product Details & Documents
Synonyms: 4-Bromo-1H-indole
- CAS Number:52488-36-5
- Formula:C₈H₆BrN
- MDL Number:MFCD00671502
- Molecular Weight:196.05 g/mol
Specifications
Specifications
- Tipo di conf.:Glass bottle for solids
- Conf:1 g



