Orders LinkContactUsLinkComponent
2,6-Dichloropyridine
Glass bottle for solids, 100 g
  COBBOS-7427-100G
Marketsource
2,6-Dichloropyridine
  COBBOS-7427-100G
Marketsource
 :  2402-78-0
2,6-Dichloropyridine
Glass bottle for solids, 100 g
 :  2402-78-0
1 * 100 g
  •  :2402-78-0
  •  :C₅H₃Cl₂N
  •  :MFCD00006244
  •  :147.99 g/mol
  •  :211 °C (1013 hPa)
  •  :83…86 °C
  •  :Ambiente
  •  :219-282-3
  •  :2811
  •  :6.1,III
  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    100 g