2,6-Dichloropyridine
Glass bottle for solids, 100 g
COBBOS-7427-100G
Marketsource
- :2402-78-0
- :C₅H₃Cl₂N
- :MFCD00006244
- :147.99 g/mol
- :211 °C (1013 hPa)
- :83…86 °C
- :Ambiente
- :219-282-3
- :2811
- :6.1,III
- Tipo di conf.:Glass bottle for solids
- Conf:100 g