2'-Bromoacetophenone
Glass bottle for solids, 100 g
COBBOS-7606-100G
Marketsource
: o-Bromoacetophenone
- :2142-69-0
- :C₈H₇BrO
- :MFCD00000067
- :199.05 g/mol
- :249 °C (1013 hPa)
- :0 °C
- :1,476 g/cm³ (20 °C)
- :Ambiente
- :218-398-1
- Tipo di conf.:Glass bottle for solids
- Conf:100 g