Orders LinkContactUsLinkComponent
2'-Bromoacetophenone
Glass bottle for solids, 100 g
  COBBOS-7606-100G
Marketsource
2'-Bromoacetophenone
  COBBOS-7606-100G
Marketsource
 :  2142-69-0
2'-Bromoacetophenone
Glass bottle for solids, 100 g
 :  2142-69-0
1 * 100 g
 :  o-Bromoacetophenone
  •  :2142-69-0
  •  :C₈H₇BrO
  •  :MFCD00000067
  •  :199.05 g/mol
  •  :249 °C (1013 hPa)
  •  :0 °C
  •  :1,476 g/cm³ (20 °C)
  •  :Ambiente
  •  :218-398-1
  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    100 g