2'-Bromoacetophenone
Glass bottle for solids, 5 g
Catalog # COBBOS-7606-5G
Marketsource
Supplier: COMBI-BLOCKS
2'-Bromoacetophenone
Catalog # COBBOS-7606-5G
Supplier: COMBI-BLOCKS
Marketsource
CAS Number: 2142-69-0
Jump to:
Product Details & Documents
Synonyms: o-Bromoacetophenone
- CAS Number:2142-69-0
- Formula:C₈H₇BrO
- MDL Number:MFCD00000067
- Molecular Weight:199.05 g/mol
- Boiling Pt:249 °C (1013 hPa)
- Melting Pt:0 °C
- Density:1,476 g/cm³ (20 °C)
- Storage Temperature:Ambiente
- EINECS:218-398-1
Specifications
Specifications
- Tipo di conf.:Glass bottle for solids
- Conf:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



