Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
5-Bromopicolinamide

5-Bromopicolinamide

Bottiglia di vetro, 1 g
Catalog # COBBCA-4207-1G
Marketsource
Supplier:   COMBI-BLOCKS
5-Bromopicolinamide
Catalog # COBBCA-4207-1G
Supplier:  COMBI-BLOCKS
Marketsource
CAS Number:  90145-48-5
5-Bromopicolinamide

Bottiglia di vetro, 1 g

CAS Number:  90145-48-5
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  5-Bromopyridine-2-carboxamide
  • CAS Number:90145-48-5
  • Formula:C₆H₅BrN₂O
  • MDL Number:MFCD04066715
  • Molecular Weight:201.0200 g/mol
Specifications

Specifications

  • Tipo di conf.:
    Bottiglia di vetro
  • Conf:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.