Razor blades, Gem®
Lame per rasoio a bordo singolo, Gem® Scientific, 3 sfaccettature, retro in alluminio, 100 lame a dispenser, Acciaio al carbonio, 0,22 mm, 38×19 mm
Codice catalogo PERS62-0325
Nome del fornitore: PERSONNA MEDICAL
Vai a
Product Details & Documents
Razor blades for catheter cutting or cleanroom applications.
- High precision, sharpness and safety
- Made from carbon steel or from stainless steel
Carbon steel blades are suitable for ISO Class 7 cleanroom environments.
Stainless steel blades are suitable for ISO Class 5, 6 and 7 cleanroom environments.
Specifiche
- Descrizione:Lame per rasoio a bordo singolo, Gem® Scientific, 3 sfaccettature, retro in alluminio, 100 lame a dispenser
- L×L:38×19 mm
- Materiale:Acciaio al carbonio
- Spessore:0,22 mm