Acrylamide/ Bis-Acrylamide
Acrylamide/bisacrylamide (29:1), premixed powder
Codice catalogo 786-507
Nome del fornitore: G-Biosciences
Vai a
Product Details & Documents
Acrylamide (electrophoresis grade) is supplied as a powder or a 40% solution in ultrapure water. Bisacrylamide (Bis (N,N’-methylenebisacrylamide)) (electrophoresis grade) is supplied as a powder or a 2% solution in ultrapure water.
Acrylamide/bisacrylamide solutions are available at the most common ratios (37,5:1 or 29:1) for use in protein and nucleic acid electrophoresis. The concentration is based on the total weight of both the acrylamide and bisacrylamide. Supplied as 40% solutions prepared from electrophoresis grade acrylamide and bis-acrylamide in ultra-pure water.
Specifiche
- Descrizione:Acrylamide/bisacrylamide (29:1), premixed powder
- Seq:08