Orders LinkContactUsLinkComponent
Acrylamide/ Bis-Acrylamide
Acrylamide/ Bis-Acrylamide
Acrylamide/bisacrylamide (37.5:1), 40% solution
Catalog # 786-502
Supplier:   G-Biosciences
Acrylamide/ Bis-Acrylamide
Catalog # 786-502
Supplier:  G-Biosciences
Acrylamide/ Bis-Acrylamide
Acrylamide/ Bis-Acrylamide
Acrylamide/bisacrylamide (37.5:1), 40% solution
Price
SFr. 236.00
Log in for availability
1 * 500 mL
Product Details & Documents
Acrylamide (electrophoresis grade) is supplied as a powder or a 40% solution in ultrapure water. Bisacrylamide (Bis (N,N’-methylenebisacrylamide)) (electrophoresis grade) is supplied as a powder or a 2% solution in ultrapure water.
Acrylamide/bisacrylamide solutions are available at the most common ratios (37,5:1 or 29:1) for use in protein and nucleic acid electrophoresis. The concentration is based on the total weight of both the acrylamide and bisacrylamide. Supplied as 40% solutions prepared from electrophoresis grade acrylamide and bis-acrylamide in ultra-pure water.
Specifications
  • Descrizione:
    Acrylamide/bisacrylamide (37.5:1), 40% solution
  • Seq:
    03