Gel drying solution for protein gels
Gel drying solution
786-686
This gel drying solution effectively and evenly dries protein electrophoresis gels by the mechanism of passive evaporation and is suitable for permanent gel storage and densitometry.
- Ready to use
- Regulates rate of drying
- Prevents cracking of the gel
- Compatible with all drying membranes
Documents
- Descrizione:Gel drying solution
- Seq:03