Orders LinkContactUsLinkComponent
Stirrer blades
Propeller with four blades, PTFE/PEEK, shaft Ø 10 mm, 200×30×5 mm, wrench 19 mm
  BOHLC448-28
 :   Bohlender
Stirrer blades
  BOHLC448-28
 :  Bohlender
Stirrer blades
Stirrer blades
Propeller with four blades, PTFE/PEEK, shaft Ø 10 mm, 200×30×5 mm, wrench 19 mm
SFr. 943.00
1 * 1 ST
Stirrer blades made from PTFE, with a set of clamp screws made from a PTFE/PEEK compound.
  • Chemical resistant
  • Temperature resistance from –200 to +250 °C
  • Create axial flow
The blades can be fixed tightly on stirrer shafts by means of the clamp screws. A spanner wrench is included for easy assembly.
  • Tipo:
    Propeller with four blades, PTFE/PEEK, shaft Ø 10 mm, 200×30×5 mm, wrench 19 mm
  • Seq:
    18