Orders LinkContactUsLinkComponent
Vario couplings
Vario couplings
Couplings with GL male thread, 8,0 mm, Connecting laboratory screw joints and elastic tubing wall thickness 1,6 and 2,4 mm, GL 18, 8 / 9,5 mm
Codice catalogo BOHLD681-48
Nome del fornitore:   Bohlender
Vario couplings
Codice catalogo BOHLD681-48
Nome del fornitore:  Bohlender
Vario couplings
Vario couplings
Couplings with GL male thread, 8,0 mm, Connecting laboratory screw joints and elastic tubing wall thickness 1,6 and 2,4 mm, GL 18, 8 / 9,5 mm
Price
SFr. 174.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Vario 2-part coupling to connect elastic tubing with non-elastic tubing made of PTFE, glass, metal or other plastics.
  • The elastic tubing is pushed on a cone and fixed on the coupling by means of a connecting nut
  • These couplings are available in two different versions, according to male and female threads to connect
Specifiche
  • Per:
    Connecting laboratory screw joints and elastic tubing wall thickness 1,6 and 2,4 mm
  • Ø foro:
    8,0 mm
  • Filettatura:
    GL 18
  • Ø int.tubo:
    8 / 9,5 mm
  • Descrizione:
    Couplings with GL male thread