Orders LinkContactUsLinkComponent
Vario couplings | Avantor
Vario couplings
Couplings with GL male thread, 1,6 mm, Connecting laboratory screw joints and elastic tubing wall thickness 0,8 and 1,6 mm, GL 14, 1,6 mm
Codice catalogo BOHLD681-16
Nome del fornitore:   Bohlender
Vario couplings
Codice catalogo BOHLD681-16
Nome del fornitore:  Bohlender
Vario couplings
Vario couplings
Couplings with GL male thread, 1,6 mm, Connecting laboratory screw joints and elastic tubing wall thickness 0,8 and 1,6 mm, GL 14, 1,6 mm
Price
SFr. 79.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Vario 2-part coupling to connect elastic tubing with non-elastic tubing made of PTFE, glass, metal or other plastics.
  • The elastic tubing is pushed on a cone and fixed on the coupling by means of a connecting nut
  • These couplings are available in two different versions, according to male and female threads to connect
Specifiche
  • Per:
    Connecting laboratory screw joints and elastic tubing wall thickness 0,8 and 1,6 mm
  • Ø foro:
    1,6 mm
  • Filettatura:
    GL 14
  • Ø int.tubo:
    1,6 mm
  • Descrizione:
    Couplings with GL male thread
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..