Orders LinkContactUsLinkComponent
Vario couplings
Vario couplings
Couplings with GL male thread, 1,6 mm, Connecting laboratory screw joints and elastic tubing wall thickness 0,8 and 1,6 mm, GL 14, 1,6 mm
  BOHLD681-16
 :   Bohlender
Vario couplings
  BOHLD681-16
 :  Bohlender
Vario couplings
Vario couplings
Couplings with GL male thread, 1,6 mm, Connecting laboratory screw joints and elastic tubing wall thickness 0,8 and 1,6 mm, GL 14, 1,6 mm
SFr. 79.00
1 * 1 ST
Vario 2-part coupling to connect elastic tubing with non-elastic tubing made of PTFE, glass, metal or other plastics.
  • The elastic tubing is pushed on a cone and fixed on the coupling by means of a connecting nut
  • These couplings are available in two different versions, according to male and female threads to connect
  • Per:
    Connecting laboratory screw joints and elastic tubing wall thickness 0,8 and 1,6 mm
  • Ø foro:
    1,6 mm
  • Filettatura:
    GL 14
  • Ø int.tubo:
    1,6 mm
  • Descrizione:
    Couplings with GL male thread