Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
O-ring seals for flat flanges, Quickfit®

O-ring seals for flat flanges, Quickfit®

Flat flanges, flange bore 100 mm, 0,5 mm
  BIBBPS100
 :   SciLabware
O-ring seals for flat flanges, Quickfit®
  BIBBPS100
O-ring seals for flat flanges, Quickfit®
O-ring seals for flat flanges, Quickfit®

Flat flanges, flange bore 100 mm, 0,5 mm

 
45.90 SFr.
   
1 * 1 ST
 

PTFE, white.

  • Excellent chemical resistance
  • Excellent temperature resistance
  • Forms tight seal with flat flange joints

For use with Quickfit® flat flange jointed glassware.

 
  • Per:
    Flat flanges, flange bore 100 mm
  • Spessore parete:
    0,5 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.