Orders LinkContactUsLinkComponent
IP lysis buffer, Pierce™
IP lysis buffer, Pierce™
IP lysis buffer, 250 ml
Codice catalogo PIER87788
Nome del fornitore:   Thermo Fisher Scientific
IP lysis buffer, Pierce™
Codice catalogo PIER87788
Nome del fornitore:  Thermo Fisher Scientific
Numero CAS:  
IP lysis buffer, Pierce™
IP lysis buffer, Pierce™
IP lysis buffer, 250 ml
Numero CAS:  
Price
392.00 SFr.
Log in per disponibilità
1 * 250 mL
Product Details & Documents
The IP lysis buffer is a mammalian whole cell lysis reagent based on a modified RIPA buffer formulation without SDS.
  • Compatible with protein assays, reporter assays and immunoassay procedures
  • Optimized for compatibility with immunoprecipitation and pull-down assays
  • Ready-made formula is effective for extracting cytoplasmic, membrane and nuclear proteins
  • Gentle formulation helps maintain protein complexes for co-immunoprecipitation
This buffer is effective for lysing cultured mammalian cells from both plated cells and cells pelleted from suspension cultures. They are composed of 25 mM Tris-HCl pH 7,4, 150 mM NaCl, 1 mM EDTA, 1% NP-40 and 5% glycerol. The buffer does not contain protease or phosphatase inhibitors.
Specifiche
Specifiche
  • Conf:
    250 ml
  • Descrizione:
    IP lysis buffer
Specifiche