Nalgene® Media Bottles, Square, with Screw Cap
30 ml, 64 mm, 14 mm, 20-415
Catalog # NALG322020-0030
Supplier: Thermo Fisher Scientific
Nalgene® Media Bottles, Square, with Screw Cap
Catalog # NALG322020-0030
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PETG bottle with HDPE screw closure.
- Graduated
- Durable
- Break-resistant
- Leakproof
Package and ship liquid media, buffers and sera using square media bottles with screw caps. Heavy-walled, durable bottles offer excellent gas barrier properties and are specifically designed for storing media and sera. Non-sterile bottle offers excellent gas barrier properties; clear PETG material allows for easy identification of contents.
Certifications: USP Class VI, FDA Compliance 21 CFR
Specifications
- Filettatura:20-415
- Capacità:30 ml
- Altezza:64 mm
- Seq:01
- Ø interno collo:14 mm
- Cat. No.:NALG322020-0030
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:NAR Certificate of Retirement
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



