Gastro-norm container
6,5 l, 325×265×100 mm
Catalog # HUPF0130782H
Supplier: HUPFER METALLWERKE
Jump to:
Product Details & Documents
Stainless steel 18/10.
- Fast, easy cleaning
- Good chemical resistance
- Very good thermal resistance
Multipurpose containers for a wide range of uses.
Specifications
- Capacità:6,5 l
- Seq:01
- Esterno L×P×A:325×265×100 mm