Autoclaves, front loading, D-Line
Tuttnauer's laboratory autoclaves are ideal for a wide range of applications in various fields, such as research institutes, universities, pharmaceutical, food, medical and biotechnological industries. This advanced laboratory line is a quality range of reliable bench top (front loading) and vertical (top loading) models with chamber volumes from 28 to 160 litres that provides a single solution for the full spectrum of sterilisation needs. The autoclaves can be fully configured from general use to customer-specific applications. Units from either the Essential or Hi End ranges can be customised with various different optional features to meet more specific or demanding applications.
- High quality AISI 316 L stainless steel chamber with mirror finished door
- Advanced control system with easy to use three-button keypad
- Up to 20 customisable cycle programs
- Standard USB and Ethernet connection allows easy transfer of cycle data to a PC
- Two flexible PT100 temperature sensors
The Essential bench top (front loading) laboratory autoclaves fit on benches or work tops. These basic stand-alone models are well known for their steam production inside the chamber by means of heating elements around the chamber. This allows the user to run basic sterilisation cycles for solid loads, liquids and waste (solid or liquid). Adding different options and accessories enables the user to achieve even more functionality from the autoclave such as quicker cooling times, dry products at the end and safe handling of hazardous loads.
Easy to clean chamber
Manual filling and draining of the demineralised water reservoir
Re-use of the demineralised water
The Hi End horizontal (front loading) laboratory autoclaves 2840 ELC-G-D model fits directly on benches or counter tops, where the 38xx and 50xx ELC-G-D models fit directly on the laboratory floor without the need for a support table. These models are designed to be connected directly to utilities and are well known for their steam production in the built-in steam generator as well as the standard fast cooling option. This allows the user to run liquid cycles with fast cooling in addition to the basic sterilisation cycles for solid loads, liquids and waste (solid or liquid). Adding different options and accessories enables the user to achieve even more functionality from the autoclave such as faster cooling times, dry products at the end and safe handling of hazardous loads.
Full utilities required (demineralised water (not for model 2840), tap water, drain connection, compressed air and electricity)
Automatic filling and draining functionality (optional for model 2840)
Automatic cooling of the exhaust to the drain 60 °C (by parameter)
Ordering information: Accessories as well as a range of factory fitted options such as units with fast/air cooling, super fast cooling, pre-/post-vacuum and models suitable for biohazardous waste sterilisation are available on request. Please contact your local Avantor sales office for further details prior to ordering as these cannot be retro-fitted.
Delivery information: Note: all 2840 and the Essential 3850 models require 230 V single phase and the Hi End 3850 and all 3870, 5050 and 5075 models require a 400 V 3 phase supply.
Documents
CoC Autoclave Models for EL-D, ELC-D, ELV-D and ELVC-D Series CoC Autoclave Models for ELCG-D and ELVCG-D Series
Manual Electronic Laboratory Autoclaves Models 2840 EL-D Manual Electronic Laboratory Autoclaves Models 2840 ELG-D Manual Electronic Laboratory Autoclaves Models 3840, 3850, 3870 EL-D
Manual
Tuttnauer Vertical and Benchtop Autoclaves for Life Sciences
Product Literature
- Capacità:28 L
- Descrizione:Autoclave, caricamento anteriore, 2840 EL-D, con pannello di controllo a microprocessore avanzato
- Seq:001
- Cat. No.:481-0555
- Pressione max.:2,8 bar
- Peso:43 kg
- Ø×A interne:280×400 mm
- Materiale:Acciaio inossidabile 316L
- Temperatura:134 °C
- Modello:2840 EL-D
- L×P×A est.:530×630×440 mm
- Potenza:2200 W
Accessories
- Per
- DescrizionePer
- 481-0851Carta per stampante termica THE-002-0052 (5 rotoli)Modelli EL-ELV e D-line1 * 5 ST - SFr. 13.90
- DescrizioneCapacitàPer
- TUTTBSK411-0003Contenitore in acciaio inox, 265L×390W mm, 21,5 L__2840 autoclave1 * 1 ST - SFr. 1’570.00
- DescrizioneCapacità
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



