Sulfonation flasks, with three necks
1500 ml, 45/40 NS, 29/32 NS
Codice catalogo WITG0.851.150
Nome del fornitore: witeg Labortechnik
Sulfonation flasks, with three necks
Codice catalogo WITG0.851.150
Nome del fornitore: witeg Labortechnik
Vai a
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With one parallel and two angled side necks.
Specifiche
- Capacità:1500 ml
- Seq:05
- Colli laterali:29/32 NS
- Collo centrale:45/40 NS
- Cat. No.:WITG0.851.150