Drum funnels
430 mm, In PE, 37 mm
Codice catalogo BURK9602-0450
Nome del fornitore: BURKLE
Vai a
Product Details & Documents
Made of PP, translucent.
- Very good chemical resistance
- Very good temperature resistance
- Quick and easy to clean
For filling containers with large quantities of liquid or powdered substances quickly.
Documents
BURK9602-0250_TDS_EN
Scheda dati
Specifiche
- Ø esterno imbuto:430 mm
- Materiale:In PE
- Seq:05
- Ø esterno stelo:37 mm