Caps
Cap kit, includes one cap, with fittings, one 1 l safety coated glass bottle, 3 m PFA tubing, one No-Met 5 µm mobile phase filter, GL 45, 1 port for 1/16" tubing, 2 ports for 1/8" and 1/16" OD tubing, PP
Codice catalogo JOURJR-S-11102
Nome del fornitore: VICI JOUR RESEARCH
Vai a
Product Details & Documents
These caps top bottles are available with two, three, or four colour-coded 1/4-28 thread for ¹/₈ or ¹/₁₆" O.Ø tubing.
Specifiche
- Filettatura:GL 45
- Descrizione:Cap kit, includes one cap, with fittings, one 1 l safety coated glass bottle, 3 m PFA tubing, one No-Met 5 µm mobile phase filter
- Materiale:PP
- Seq:14
- Connettore:1 port for 1/16" tubing, 2 ports for 1/8" and 1/16" OD tubing
- Cat. No.:JOURJR-S-11102